Fortfahren mit
MTCP4408
MTC™ Bio SureStand™ Multiple Pipette Stand
to hold, 8 x single pipettes OR 6 x multipipettes
Synonym(e):
pipette holder, MTC Bio P4408
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
NACRES:
NB.02
UNSPSC Code:
41122801
eCl@ss:
32032209
Manufacturer/tradename:
MTC™ Bio P4408
Feature:
to hold 8 x single pipettes OR 6 x multipipettes
Material:
acrylic
Packaging:
pack of 1 ea
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
acrylic
feature
to hold 8 x single pipettes OR 6 x multipipettes
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio P4408
General description
SureStand™ Pipette Stand for 8 pipettes, up to six multi-channels, acrylic
SureStand™ Pipette racks are unique in their ability to accommodate multi-channel pipettes without sacrificing any of the stability of a traditional acrylic rack. The patented “windows” allow the user to insert the multi-channel head longways through the rack while keeping the body of the pipette stable by securing the pipette at two points: under the finger hook and near the bottom of the handle. Each SureStand™ is also outfitted with four non-skid rubber “feet” for extra stability. SureStand™ is guaranteed to be the sturdiest and most versatile pipette rack in your lab.
Features and Benefits
  • Universal – Fits all popular pipettes
  • 5mm acrylic – 40% thicker
  • Non-skid rubber feet
  • Multi-channel “window” slots
  • US Patent No 872,300
Legal Information
MTC is a trademark of MTC Bio
SureStand is a trademark of MTC Bio
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)