Fortfahren mit
MTCR8013
MTC™ Bio IceRack™ Tube Platform with Detachable Lid
to hold, 72 x 1.5 mL tubes
Synonym(e):
cooled microtube rack, cooled tube rack, benchtop cold tube holder, cooled tube holder, MTC Bio T8013
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
NACRES:
NB.12
UNSPSC Code:
41122806
eCl@ss:
32032201
Manufacturer/tradename:
MTC™ Bio R8013
Feature:
to hold 72 x 1.5 mL tubes
Material:
polypropylene
Packaging:
pack of 6 ea
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
polypropylene
feature
to hold 72 x 1.5 mL tubes
packaging
pack of 6 ea
manufacturer/tradename
MTC™ Bio R8013
parameter
-80-135 °C temp. range
General description
IceRack™ boxes have a removable white tube tray that allows the blue base to be filled with crushed ice or other temperature-controlling materials. They also feature a translucent cover. Temperature-sensitive samples and assays in microtubes can be processed with an extra degree of temperature stability. These boxes are suitable for use with screw-cap or snap-cap microtubes from 0.5mL to 1.5/2.0mL.
IceRack™ for 72 × 1.5mL tubes, detachable lid, tube platform, and base, numbered cavities, accepts snap cap and screw cap 1.5mL to 2.0mL tubes, 6/cs
Features and Benefits
  • Designed for crushed Ice
  • Polypropylene, -80°C to 135°C
  • Detachable “lift-off” lids
  • Economical
  • For screw cap and snap cap tubes
Legal Information
IceRack is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)