Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 pkg | Warenkorb auf Verfügbarkeit prüfen | € 127,00 |
Über diesen Artikel
€ 127,00
material
glass
packaging
pack of 100 ea
manufacturer/tradename
Drummond Scientific 5-000-1001-X
technique(s)
gas chromatography (GC): suitable using :Suitable
capacity
1 - 5 μL
volume range
1-5 μL
General description
Application
Packaging
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description volume range 1-5 μL | description volume range 1-10 μL | description capacity 1-5 μL | description capacity 100 μL |
| manufacturer/tradename Drummond Scientific 5-000-1001-X | manufacturer/tradename Drummond Scientific 5-000-1001-X10 | manufacturer/tradename Drummond Scientific 2-000-001 | manufacturer/tradename Drummond Scientific 2-000-100 |
| material glass | material glass | material glass | material glass |
| packaging pack of 100 ea | packaging pack of 100 ea | packaging pack of 250 ea, pack of 1 | packaging pack of 250 ea |
| technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) - | technique(s) - |
| capacity 1 - 5 μL | capacity 10 μL | capacity 1-5 μL | capacity 100 μL |
| volume range 1-5 μL | volume range 1-10 μL | volume range - | volume range - |
Still not finding the right product?
Explore all of our products under Microcapillary pipettes, positive displacement
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?