Fortfahren mit
Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
biological source
yeast
Quality Segment
sterility
0.2 μm filtered
product line
BioReagent
form
liquid
shelf life
2 yr
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
-10 to -25°C
General description
Application
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description 0.2 μm filtered, BioReagent, suitable for cell culture | description Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture | description with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture | description Solution Stabilized, with 5,000 units penicillin and 5mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture |
| technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| product line BioReagent | product line BioReagent | product line BioReagent | product line BioReagent |
| form liquid | form liquid | form liquid | form liquid |
| storage temp. -10 to -25°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| biological source yeast | biological source - | biological source - | biological source - |
Lagerklasse
12 - Non Combustible Liquids
WGK
WGK 2
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

