Direkt zum Inhalt
Merck

Fortfahren mit

PLA0095

Rabbit anti-SRC3 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(e):

RAC-3, ACTR, AIB-1, AIB1, CAGH16, CBP-interacting protein, CTG26, KAT13B, NCoA-3, RAC3, SRC-3, SRC3, TNRC14, TNRC16, TRAM-1, amplified in breast cancer 1 protein, bHLHe42, class E basic helix-loop-helix protein 42, p/CIP, pCIP, receptor-associated coactivator 3, steroid receptor coactivator protein 3, thyroid hormone receptor activator molecule 1

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Über diesen Artikel

NACRES:
NA.41
UNSPSC Code:
12352203
Species reactivity:
human
Technique(s):
immunoprecipitation (IP): 2-10 μg/mg, western blot: 1:2,000-1:10,000
Application:
IP, WB
Citations:
2


biological source

rabbit

Quality Segment

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-10 μg/mg, western blot: 1:2,000-1:10,000

accession no.

NP_858045.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... SRC3(8202)

General description

Steroid receptor coactivator-3 (SRC-3) is a transcriptional coactivator,. It belongs to the p160 SRC family. SRC-3 is located on human chromosome 20q13.

Immunogen

The epitope recognized by PLA0095 maps to a region between residue 900 and 950 of human Steroid Receptor Coactivator 3 using the numbering given in entry NP_858045.1 (GeneID 8202).

Biochem/physiol Actions

Steroid receptor coactivator-3 (SRC-3) participates in the genesis and development of various cancers. SRC-3 possesses histone acetyltransferase activity. It modulates the expression of target genes, that participates in various biological processes, such as cell proliferation, differentiation and migration. SRC-3 increases the migration and invasion of esophageal squamous cell carcinoma (ESCC) cells.

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

Steroid receptor coactivator 3 (SRC3) belongs to the SRC family of proteins that interacts with nuclear receptors to facilitate transcription. The 3 homologous members of this family are SRC-1 (NCoA-1), SRC-2 (GRIP1, TIF2, or NcoA-2), and SRC-3 (p/CIP, RAC3, ACTR, AIB1, or TRAM-1). SRC3 is an oncogene that is reported to be amplified and/or overexpressed in breast and gastric cancer and stimulates AKT signaling and cell growth and tumorigenesis.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Probieren Sie unser Produkt-Auswahlhilfe, um Ihre Auswahl einzugrenzen


Lagerklasse

12 - Non Combustible Liquids

Was ist los?

nwg

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable


Zulassungslistungen

Zulassungslistungen werden hauptsächlich für chemische Produkte erstellt. Für nicht-chemische Produkte können hier nur begrenzte Angaben gemacht werden. Kein Eintrag bedeutet, dass keine der Komponenten gelistet ist. Es liegt in der Verantwortung des Benutzers, die sichere und legale Verwendung des Produkts zu gewährleisten.

77-92-9

CAS No.


Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen




Questions

Reviews

No rating value

Active Filters