Direkt zum Inhalt
Merck

Fortfahren mit

PLA0098

Rabbit anti-BIRC6/Apollon Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(e):

BIR repeat-containing ubiquitin-conjugating enzyme, BRUCE, apollon, baculoviral IAP repeat-containing 6, ubiquitin-conjugating BIR domain enzyme apollon, ubiquitin-conjugating BIR-domain enzyme apollon

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis
100 μL
Warenkorb auf Verfügbarkeit prüfen
€ 417,00

Über diesen Artikel

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
-

€ 417,00


Warenkorb auf Verfügbarkeit prüfen

​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry: 1:2,000- 1:10,000, immunoprecipitation (IP): 2-10 μg/mg, western blot: 1:2,000-1:10,000

accession no.

Q9NR09

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

Immunogen

The epitope recognized by PLA0098 maps to a region between residue 4775 and the C-terminus (residue 4829) of human Baculoviral IAP repeat-containing protein 6 using the numbering given in Swiss-Prot entry Q9NR09 (GeneID 57448).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

BIRC6/Apollon contains a baculoviral-IAP repeat (BIR) and ubiquitin-conjugating enzyme (UBC) domain and functions in the inhibition of apoptosis. BIRC6/Apollon inhibits apoptosis by ubiquitinating and promoting the proteasomal degradation of SMAC (second mitochondria-derived activator of caspase) and caspase-9.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
HPA074738PLA0248PLA0227
description

Powered by Bethyl Laboratories, Inc.

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

description

Powered by Bethyl Laboratories, Inc.

description

Powered by Bethyl Laboratories, Inc.

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

human, mouse

species reactivity

human

species reactivity

human

species reactivity

human, mouse

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

Probieren Sie unser Produkt-Auswahlhilfe, um Ihre Auswahl einzugrenzen


Lagerklasse

12 - Non Combustible Liquids

Was ist los?

nwg

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable


Zulassungslistungen

Zulassungslistungen werden hauptsächlich für chemische Produkte erstellt. Für nicht-chemische Produkte können hier nur begrenzte Angaben gemacht werden. Kein Eintrag bedeutet, dass keine der Komponenten gelistet ist. Es liegt in der Verantwortung des Benutzers, die sichere und legale Verwendung des Produkts zu gewährleisten.

77-92-9

CAS No.


Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen