Direkt zum Inhalt
Merck

Fortfahren mit

SML4191

TAMRA-Azide-Biotin

≥93% (HPLC), powder, click chemistry tool

Synonym(e):

9-[4-6-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]-26-[(3aS,4S,6aR)-hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1,5,9,23-tetraoxo-13,16,19-trioxa-2,6,10,22-tetraazahexacos-1-yl]-2-carboxyphenyl]-3,6-bis(dimethylamino)-xanthylium, TAMRA-Biotin-Azide, Azide-TAMRA-Biotin, TAMRA-Azide-PEG-Biotin, TAMRA-Azide-PEG3-Biotin, 4/5-Carboxytetramethylrhodamine-Azide-PEG3-Biotin, AzTB

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

Empirische Formel (Hill-System):
C57H79N11O14S
CAS-Nummer:
Molekulargewicht:
1174.37
Assay:
≥93% (HPLC)
Form:
powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


Quality Segment

assay

≥93% (HPLC)

form

powder

color

pink to dark red

solubility

DMSO: 2 mg/mL, clear

shipped in

dry ice

storage temp.

−70°C

Application

TAMRA-Azide-Biotin is suitable for fluorescent visualization (TAMRA fluorophore) and biochemical enrichment (biotin group). It may be used for site-specific labeling of RNA through copper-catalyzed click chemistry, enabling the visualization and isolation of RNA molecules in various biochemical assays.

Biochem/physiol Actions

A multifunctional azide-TAMRA-biotin as a proteome-wide tool; Trifunctional click chemistry probe that contains a ligation handle, a biotin and a fluorophore. The trifunctional TAMRA-Azide-Biotin capture reagent combines three functional elements: an azide group for selective reaction, TAMRA for fluorescence-based visualization, and biotin for efficient isolation. The azide group targets alkyne-modified molecules within cells, enabling specific binding to desired molecular targets. TAMRA illuminates these targets, allowing researchers to track their location and concentration, thus shedding light on cellular dynamics. Biotin, as the binding affinity molecule, facilitates the subsequent isolation or analysis of these targets, especially when used with streptavidin or avidin-coated surfaces. The hydrophilic PEG spacer enhances solubility in aqueous solutions. This integration of reaction specificity, visualization, and isolation capabilities makes TAMRA-Azide-Biotin invaluable for experiments that require detailed detection and analysis of specific cellular components, such as protein interaction studies or real-time monitoring of cellular processes. Abs/Em ~ 545/565 nm.


Lagerklasse

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen