Fortfahren mit
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenbiological source
fermentation/recombinant (ACF)
Quality Segment
form
liquid
shelf life
2 yr (Retest)
mol wt
17.6 kDa
concentration
0.4-0.6 mg/mL
storage temp.
-10 to -25°C
General description
Fibroblast Growth Factor-2 (FGF2) also known as basic FGF (bFGF) is a heparin-binding growth factor that stimulates the proliferation of a wide variety of cells including mesenchymal, neuroectodermal and endothelial cells. FGF2 also exerts a potent angiogenic activity in vivo. It synergizes with the BMP antagonist noggin to sustain undifferentiated proliferation of human embryonic stem (hES) cells under feeder-free conditions. Due to its rapid degradation of wild-type FGF2 at 37 °C, stem cell cultures require frequent media changes with FGF2 supplementation to support continual cell prolifieration. StableCell™ FGF-2 is an engineered version of human FGF2 exhibiting enchanced thermostability. StableCell™ FGF2 enables a more relaxed feeding schedule to support human ES/iPS and neural stem cell culture.
Biochem/physiol Actions
StableCell™ FGF-2 is a proprietary engineered protein sequence under license from Enantis. It is expressed in E. coli and is animal component free. StableCell™ FGF-2 is engineered with enchanced thermostability compared to the wild-type bFGF protein.
Physical form
StableCell™ FGF2 is supplied as a 0.2 µm filtered aqueous solution in 20 mM phosphate buffer pH 7.5, 500 mM NaCl.
Legal Information
StableCell is a trademark of Sigma-Aldrich Co. LLC
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description Animal-component free, aqueous solution 0.4-0.6 mg/ml, suitable for cell culture | description FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture | description - | description 1X, sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red |
| Quality Level 300 | Quality Level 200 | Quality Level 100 | Quality Level 500 |
| biological source fermentation/recombinant (ACF) | biological source human | biological source rabbit | biological source - |
| shelf life 2 yr (Retest) | shelf life - | shelf life - | shelf life - |
| concentration 0.4-0.6 mg/mL | concentration - | concentration 1.1 mg/mL | concentration 1X |
| storage temp. -10 to -25°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
| form liquid | form lyophilized powder | form liquid | form - |
Lagerklasse
10 - Combustible liquids
WGK
WGK 1
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.



