Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
borosilicate glass 3.3 , clear bottle, pink polypropylene cap, wide-mouth
joint
joint threaded neck
feature
with cap
manufacturer/tradename
Kemtech B141000
technique(s)
cell culture: suitable
capacity
500 mL
fitting
GL 45 bottle
General description
Application
Features and Benefits
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description borosilicate glass 3.3, capacity 500 mL, pink polypropylene cap | description borosilicate glass 3.3, capacity 1000 mL, red polypropylene cap | description capacity 50 mL, stopper size #3 | description borosilicate glass 3.3, capacity 50 mL |
| feature with cap | feature with cap | feature - | feature - |
| capacity 500 mL | capacity 1000 mL | capacity 50 mL | capacity 50 mL |
| fitting GL 45 bottle | fitting - | fitting - | fitting - |
| material pink polypropylene cap, borosilicate glass 3.3 , clear bottle, wide-mouth | material red polypropylene cap, borosilicate glass 3.3 , clear bottle, wide-mouth | material glass flask, flat bottom flask | material borosilicate glass 3.3 |
| manufacturer/tradename Kemtech B141000 | manufacturer/tradename Kemtech B140100 | manufacturer/tradename Synthware F150050 | manufacturer/tradename Kemtech S371207 |
| joint joint threaded neck | joint - | joint - | joint - |
Still not finding the right product?
Explore all of our products under Synthware™ Glass wide mouth media bottle
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?