Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 57,30 |
Über diesen Artikel
€ 57,30
material
PTFE-coated, anti-magnetic stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Ted Pella 5320
size
120 mm
General description
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description Style #3, very sharp, PTFE-coated, anti-magnetic stainless steel | description Style #5, Fine needle-sharp, PTFE-coated, anti-magnetic stainless steel | description Style #00, sharp, anti-magnetic stainless steel | description Style #5, Fine needle-sharp, anti-magnetic stainless steel |
| material PTFE-coated, anti-magnetic stainless steel | material PTFE-coated, anti-magnetic stainless steel | material anti-magnetic stainless steel | material anti-magnetic stainless steel |
| manufacturer/tradename Ted Pella 5320 | manufacturer/tradename Ted Pella 5622-TEF | manufacturer/tradename Ted Pella 5312 | manufacturer/tradename Ted Pella 505-NM |
| size 120 mm | size 110 mm | size 115 mm | size 110 mm |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Tweezers
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?