Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
12 ea | Warenkorb auf Verfügbarkeit prüfen | € 235,00 |
Über diesen Artikel
€ 235,00
material
low-density polyethylene , narrow-mouth
packaging
case of 12 ea
manufacturer/tradename
Nalgene 2425-1002
bottle volume
1000 mL
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description low-density polyethylene, bottle volume 1000 mL, ethyl alcohol, case of 12 ea | description low-density polyethylene, bottle volume 1000 mL, isopropanol, case of 12 ea | description low-density polyethylene, bottle volume 1000 mL, methanol, case of 12 ea | description low-density polyethylene, bottle volume 1000 mL, distilled water, case of 12 ea |
| manufacturer/tradename Nalgene 2425-1002 | manufacturer/tradename Nalgene 2425-1004 | manufacturer/tradename Nalgene 2425-1003 | manufacturer/tradename Nalgene 2425-1005 |
| material narrow-mouth , low-density polyethylene | material low-density polyethylene , narrow-mouth | material narrow-mouth , low-density polyethylene | material low-density polyethylene , narrow-mouth |
| packaging case of 12 ea | packaging case of 12 ea | packaging case of 12 ea | packaging case of 12 ea |
| bottle volume 1000 mL | bottle volume 1000 mL | bottle volume 1000 mL | bottle volume 1000 mL |
Still not finding the right product?
Explore all of our products under Nalgene® narrow-mouth Right-to-Know wash bottle
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?