Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
clear linear low-density polyethylene (LLDPE)
sterility
sterile
packaging
pkg of 2000 ea (2 boxes of 1000EA)
manufacturer/tradename
Whirl-Pak® B01421
bag W × L × thickness
19 cm × 30 cm × 0.076 mm
capacity
1627 mL (55 oz)
General description
Features and Benefits
- Perforated for easy opening and to protect sterility
- Designed for quick testing and disposal when the sample does not have to be retained
- Bag can be frozen to any temperature, even with liquid nitrogen, but should not be used at temperatures above 180°F (82°C)
- FDA and USDA Approved
- Post-manufacturing sterilization
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description capacity 1627 mL (55 oz), bag W × L × thickness 19 cm × 30 cm × 0.076 mm | description capacity 5441 mL (184 oz), bag W × L × thickness 37.5 cm × 50 cm × 0.102 mm | description capacity 1627 mL (17 oz), W × L × thickness 19 cm × 30 cm × 0.064 mm | description 1242 mL capacity(42 oz) |
| capacity 1627 mL (55 oz) | capacity 5441 mL (184 oz) | capacity 1627 mL (17 oz) | capacity 1242 mL capacity(42 oz) |
| material clear linear low-density polyethylene (LLDPE) | material clear linear low-density polyethylene (LLDPE) | material black linear low-density polyethylene (LLDPE) | material - |
| packaging pkg of 2000 ea (2 boxes of 1000EA) | packaging pkg of 500 ea (2 boxes of 250EA) | packaging pkg of 1000 ea (2 boxes of 500EA) | packaging pkg of 500 ea |
| bag W × L × thickness 19 cm × 30 cm × 0.076 mm | bag W × L × thickness 37.5 cm × 50 cm × 0.102 mm | bag W × L × thickness - | bag W × L × thickness - |
| manufacturer/tradename Whirl-Pak® B01421 | manufacturer/tradename Whirl-Pak® B01474 | manufacturer/tradename Whirl-Pak® Whirl-pak® B01558 | manufacturer/tradename Whirl-Pak® B01027WA |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
Still not finding the right product?
Explore all of our products under Whirl-Pak® Blender Bag
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?