Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 377,00 |
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41100000
eCl@ss:
32100300
Manufacturer/tradename:
IBI ACCRL0002
Material:
acrylic platform, aluminum support rod
€ 377,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
acrylic platform, aluminum support rod
manufacturer/tradename
IBI ACCRL0002
W × D × H
18 in. × 16 in. × 18 3/4 in.
General description
Product contains two each of the raised platforms that will accommodate two additional Low Profile Roller. This raised platform is designed to fit inside an incubator to allow placement of three Low Profile rollers inside.
Constructed of cast acrylic and aluminum for years of service inside high humidity, CO2 incubators without discoloring or rusting. Platforms bolt together for secure placement of Low Profile rollers.
Constructed of cast acrylic and aluminum for years of service inside high humidity, CO2 incubators without discoloring or rusting. Platforms bolt together for secure placement of Low Profile rollers.
Features and Benefits
- Allows placement of two additional rollers inside incubator
- Constructed of cast acrylic and aluminum
- Save lab counter space by stacking three rollers.
- Platforms bolt together for secure placement of Low Profile rollers
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description - | description Lab Start-Up Kit | description extra roller | description low-profile roller, AC/DC input 230 V AC, European 2-pin plug |
| manufacturer/tradename IBI ACCRL0002 | manufacturer/tradename IBI ROLLS0001 | manufacturer/tradename IBI ACCRLSR10 | manufacturer/tradename IBI |
| material acrylic platform, aluminum support rod | material - | material - | material - |
| W × D × H 18 in. × 16 in. × 18 3/4 in. | W × D × H - | W × D × H - | W × D × H - |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?