This valve is 1.03 inches in length and 0.90 inches in width, as shown in the image below:
Fortfahren mit
Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
PTFE Mininert valve
packaging
pkg of 1 ea
manufacturer/tradename
654051, VICI Precision Sampling
compatibility
for use with Hamilton TLL syringes
General description
Application
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description for use with Hamilton TLL syringes | description for use with VICI Series C and D Pressure-Lok syringes | description Luer Lock with Luer Tip, volume 50-20000 μL, for use with any Luer lock syringe | description for use with 24/400 mm thread, pkg of 12 ea |
| material PTFE Mininert valve | material PTFE Mininert valve | material stainless steel body | material white PTFE valve |
| packaging pkg of 1 ea | packaging pkg of 1 ea, 1 ea of | packaging pkg of 1 ea | packaging pkg of 12 ea |
| manufacturer/tradename 654051, VICI Precision Sampling | manufacturer/tradename 654050, VICI Precision Sampling | manufacturer/tradename SGE 031915 | manufacturer/tradename - |
| compatibility for use with Hamilton TLL syringes | compatibility for use with VICI Series C and D Pressure-Lok syringes | compatibility for use with any Luer lock syringe | compatibility for use with 24/400 mm thread |
Still not finding the right product?
Explore all of our products under Druckknopf-Ventil
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
-
What are the dimensions of this product?
1 answer