Fortfahren mit
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
NACRES:
NA.24
UNSPSC Code:
12352200
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmanufacturer/tradename
BOTANICERT®
application(s)
food and beverages
pharmaceutical
format
matrix material
storage temp.
2-25°C
General description
English name: Java Tea, French name: Ortosiphon
This botanical reference material (BRM) is manufactured by BotaniCERT™ and EXTRASYNTHESE™. The material is sourced from botanical gardens, collected by botanical experts during the flowering period, analyzed by seasoned phytochemists and geneticians.
BotaniCERT Botanical Reference Materials are samples of plants or plant parts (leaf, root, rhizome, fruit, flower, seed...) authenticated by organoleptic criteria, macroscopic and microscopic examination, HP-TLC and HPLC phytochemical fingerprint and genetic markers by PCR. They are given with 2 or 3 markers (ITS, matK, rbcL) and DNA sequences are reported.
The products are provided as 5g samples and a detailed Certificate of Analysis, including related pictures, chromatograms and DNA sequences is available.
Using a Botanical Reference Material in an identification analysis allows calibration of methods and increases the guarantee of the result obtained.
This botanical reference material (BRM) is manufactured by BotaniCERT™ and EXTRASYNTHESE™. The material is sourced from botanical gardens, collected by botanical experts during the flowering period, analyzed by seasoned phytochemists and geneticians.
BotaniCERT Botanical Reference Materials are samples of plants or plant parts (leaf, root, rhizome, fruit, flower, seed...) authenticated by organoleptic criteria, macroscopic and microscopic examination, HP-TLC and HPLC phytochemical fingerprint and genetic markers by PCR. They are given with 2 or 3 markers (ITS, matK, rbcL) and DNA sequences are reported.
The products are provided as 5g samples and a detailed Certificate of Analysis, including related pictures, chromatograms and DNA sequences is available.
Using a Botanical Reference Material in an identification analysis allows calibration of methods and increases the guarantee of the result obtained.
Application
For identity and authenticity testing of botanical materials
Legal Information
BOTANICERT is a registered trademark of BOTANICERT, Société, FR
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description Botanical reference material | description Botanicalreference material | description Botanical reference material | description Botanical reference material |
| format matrix material | format matrix material | format matrix material | format matrix material |
| application(s) food and beverages | application(s) food and beverages | application(s) food and beverages | application(s) food and beverages |
| storage temp. 2-25°C | storage temp. 2-25°C | storage temp. 2-25°C | storage temp. 2-25°C |
| manufacturer/tradename BOTANICERT® | manufacturer/tradename BOTANICERT® | manufacturer/tradename BOTANICERT® | manufacturer/tradename BOTANICERT® |
Lagerklasse
11 - Combustible Solids
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.



