Direkt zum Inhalt
Merck

Fortfahren mit

1042656

USP

Aripiprazole Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(e):

1-(2,3-Dichlorphenyl)piperazin-Hydrochlorid -hydrochlorid, 2,3-DCPP HCl

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Über diesen Artikel

Lineare Formel:
C10H12Cl2N2 · HCl
CAS-Nummer:
Molekulargewicht:
267.58
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
eCl@ss:
25140904


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

mp

243-247 °C

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].Clc1cccc(N2CCNCC2)c1Cl

InChI

1S/C10H12Cl2N2.ClH/c11-8-2-1-3-9(10(8)12)14-6-4-13-5-7-14;/h1-3,13H,4-7H2;1H

InChI key

CYQFNNSFAGXCEC-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here


Lagerklasse

11 - Combustible Solids

WGK

WGK 3

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen



Questions

Reviews

No rating value

Active Filters