Fortfahren mit
Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
grade
pharmaceutical primary standard
agency
USP
API family
colistimethate
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
-10 to -25°C
General description
Analysis Note
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) pharmaceutical (small molecule) | application(s) USP Biologics |
| format neat | format neat | format neat | format neat |
| agency USP | agency USP | agency USP | agency - |
| API family colistimethate | API family colistimethate | API family dicylomine | API family colistimethate |
Lagerklasse
11 - Combustible Solids
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.
