Direkt zum Inhalt
Merck
SCF human Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

SCF human

SCF human Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Synonym(e):
Steel Factor, Stem Cell Factor, c-Kit Ligand, Mast Cell Growth Factor (MGF)

Products

Produktkategorie
Marke
Biologische Quelle
Farbe
Physikalische Form
Reinheit
Verfahren
Showing 1-1 of 1