Merck
  • Delivery of SAR 1118 to the retina via ophthalmic drops and its effectiveness in a rat streptozotocin (STZ) model of diabetic retinopathy (DR).
Delivery of SAR 1118 to the retina via ophthalmic drops and its effectiveness in a rat streptozotocin (STZ) model of diabetic retinopathy (DR).
Investigative ophthalmology & visual science (2010-05-07)
Vidhya R Rao, Elizabeth Prescott, Namdev B Shelke, Ruchit Trivedi, Peter Thomas, Craig Struble, Tom Gadek, Charles A O'Neill, Uday B Kompella
ZUSAMMENFASSUNG
To determine the pharmacokinetics of SAR 1118, a small-molecule antagonist of leukocyte function-associated antigen (LFA)-1, after administration of ophthalmic drops in normal rats, and to determine its pharmacologic activity by assessing the inhibition of retinal leukostasis and vascular leakiness in a streptozotocin (STZ)-induced diabetic retinopathy model. The ocular pharmacokinetics of SAR 1118 were studied in rats after a single topical dose of (14)C-SAR 1118 (1 mg/eye; 40 μCi; 15.5 μL). SAR 1118 concentration time profiles in plasma and ocular tissues were quantified by liquid scintillation counting (LSC). The pharmacologic activity of SAR 1118 eye drops administered thrice daily for 2 months at 1% (0.3 mg/eye/d) and 5% (1.5 mg/eye/d) was assessed in an STZ-induced diabetic rat model by determining retinal leukostasis and blood-retinal barrier breakdown. Diabetic rats treated with periocularly administered celecoxib microparticles served as the positive control, and vehicle-treated rats served as the negative control. A single dose of 6.5% (14)C-radiolabeled SAR 1118 ophthalmic drops delivered retinal drug levels greater than 1 μM in less than 30 minutes and sustained levels greater than 100 nM for 8 hours. SAR 1118 eye drops significantly reduced leukostasis and blood-retinal barrier breakdown in a dose-dependent manner. SAR 1118 ophthalmic drops administered thrice daily deliver therapeutic levels of SAR 1118 in the retina and can alleviate the retinal complications associated with diabetes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder