Merck
  • The frequency of occurrence and discriminatory power of compounds found in human scent across a population determined by SPME-GC/MS.
The frequency of occurrence and discriminatory power of compounds found in human scent across a population determined by SPME-GC/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2006-10-03)
Allison M Curran, Carlos F Ramirez, Adee A Schoon, Kenneth G Furton
ZUSAMMENFASSUNG
The composition of human scent collected from the hands is of interest to the medical community as a mechanism to diagnose disease and the forensic community as a means to investigate canine scent discriminations. An extensive survey of the volatile organic compounds (VOCs) identified in the headspace of hand odor samples utilizing solid phase micro-extraction gas chromatography/mass spectrometry (SPME-GC/MS) has been conducted to determine the constituents of the human base odor profile. Sixty-three compounds were extracted from the collected odor samples. The composition included acids, alcohols, aldehydes, hydrocarbons, esters, ketones and nitrogen-containing compounds. The majority of the compounds detected (79.4%) were present in less than one third of the individuals sampled. Spearman correlation coefficient comparisons at a match/no-match threshold of 0.9 produced a distinguish ability of 99.67% across the population.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, light blue hub
Supelco
SPME-Faseranordnung Polyacrylat (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
Tragbarer SPME-Field-Sampler, coating PDMS/DVB
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME-Faseranordnung Polyacrylat (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
Tragbarer SPME-Field-Sampler, coating PDMS
Supelco
SPME StableFlex Methodenentwicklungs-Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, black hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME StableFlex Methodenentwicklungs-Kit, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
Tragbarer SPME-Field-Sampler, coating CAR/PDMS
Supelco
SPME-Fasersortiment-Kit 4, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Supelco
SPME-Faseranordnung, Carbowax-Polyethylenglycol (PEG)-Beschichtung, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub