Merck
  • Coupling of Grb2 to Gab1 mediates hepatocyte growth factor-induced high intensity ERK signal required for inhibition of HepG2 hepatoma cell proliferation.
Coupling of Grb2 to Gab1 mediates hepatocyte growth factor-induced high intensity ERK signal required for inhibition of HepG2 hepatoma cell proliferation.
The Journal of biological chemistry (2007-11-16)
Asuka Kondo, Naoki Hirayama, Yasuko Sugito, Michihiro Shono, Toshiaki Tanaka, Naomi Kitamura
ZUSAMMENFASSUNG
Activation of the extracellular signal-regulated kinase (ERK) pathway is a key factor in the regulation of cell proliferation by growth factors. Hepatocyte growth factor (HGF)-induced cell cycle arrest in the human hepatocellular carcinoma cell line HepG2 requires strong activation of the ERK pathway. In this study, we investigated the molecular mechanism of the activation. We constructed a chimeric receptor composed of the extracellular domain of the NGF receptor and the cytoplasmic domain of the HGF receptor (c-Met) and introduced a point mutation (N1358H) into the chimeric receptor, which specifically abrogates the direct binding of Grb2 to c-Met. The mutant chimeric receptor failed to mediate the strong activation of ERK, up-regulation of the expression of a Cdk inhibitor p16(INK4a) and inhibition of HepG2 cell proliferation by ligand stimulation. Moreover, the mutant receptor did not induce tyrosine phosphorylation of the docking protein Gab1. Knockdown of Gab1 using siRNA suppressed the HGF-induced strong activation of ERK and inhibition of HepG2 cell proliferation. These results suggest that coupling of Grb2 to Gab1 mediates the HGF-induced strong activation of the ERK pathway, which is required for the inhibition of HepG2 cell proliferation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-TrkA-Antikörper, Upstate®, from rabbit