Merck
  • ERα promotes murine hematopoietic regeneration through the Ire1α-mediated unfolded protein response.
ERα promotes murine hematopoietic regeneration through the Ire1α-mediated unfolded protein response.
eLife (2018-02-17)
Richard H Chapple, Tianyuan Hu, Yu-Jung Tseng, Lu Liu, Ayumi Kitano, Victor Luu, Kevin A Hoegenauer, Takao Iwawaki, Qing Li, Daisuke Nakada
ZUSAMMENFASSUNG
Activation of the unfolded protein response (UPR) sustains protein homeostasis (proteostasis) and plays a fundamental role in tissue maintenance and longevity of organisms. Long-range control of UPR activation has been demonstrated in invertebrates, but such mechanisms in mammals remain elusive. Here, we show that the female sex hormone estrogen regulates the UPR in hematopoietic stem cells (HSCs). Estrogen treatment increases the capacity of HSCs to regenerate the hematopoietic system upon transplantation and accelerates regeneration after irradiation. We found that estrogen signals through estrogen receptor α (ERα) expressed in hematopoietic cells to activate the protective Ire1α-Xbp1 branch of the UPR. Further, ERα-mediated activation of the Ire1α-Xbp1 pathway confers HSCs with resistance against proteotoxic stress and promotes regeneration. Our findings reveal a systemic mechanism through which HSC function is augmented for hematopoietic regeneration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Puromycin-Antikörper, Klon 12D10, clone 12D10, from mouse
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture