Merck
  • Structure of Radical-Induced Cell Death1 Hub Domain Reveals a Common αα-Scaffold for Disorder in Transcriptional Networks.
Structure of Radical-Induced Cell Death1 Hub Domain Reveals a Common αα-Scaffold for Disorder in Transcriptional Networks.
Structure (London, England : 1993) (2018-04-17)
Katrine Bugge, Lasse Staby, Katherine R Kemplen, Charlotte O'Shea, Sidsel K Bendsen, Mikael K Jensen, Johan G Olsen, Karen Skriver, Birthe B Kragelund
ZUSAMMENFASSUNG
Communication within cells relies on a few protein nodes called hubs, which organize vast interactomes with many partners. Frequently, hub proteins are intrinsically disordered conferring multi-specificity and dynamic communication. Conversely, folded hub proteins may organize networks using disordered partners. In this work, the structure of the RST domain, a unique folded hub, is solved by nuclear magnetic resonance spectroscopy and small-angle X-ray scattering, and its complex with a region of the transcription factor DREB2A is provided through data-driven HADDOCK modeling and mutagenesis analysis. The RST fold is unique, but similar structures are identified in the PAH (paired amphipathic helix), TAFH (TATA-box-associated factor homology), and NCBD (nuclear coactivator binding domain) domains. We designate them as a group the αα hubs, as they share an αα-hairpin super-secondary motif, which serves as an organizing platform for malleable helices of varying topology. This allows for partner adaptation, exclusion, and selection. Our findings provide valuable insights into structural features enabling signaling fidelity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenylmethansulfonylfluorid -Lösung, ~0.1 M in ethanol (T)
Sigma-Aldrich
Rosetta 2(DE3)-kompetente Zellen - Novagen, Novagen′s Rosetta 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.
Sigma-Aldrich
N-Octanoyl-N-Methylglucamin, ≥97% (GC)
Sigma-Aldrich
Anti-Phospho-ATM(Ser1981)-Antikörper, Klon 10H11.E12, clone 10H11.E12, Upstate®, from mouse