Merck
Targeting G-quadruplex DNA as cognitive function therapy for ATR-X syndrome.
Nature medicine (2018-05-23)
Norifumi Shioda, Yasushi Yabuki, Kouya Yamaguchi, Misaki Onozato, Yue Li, Kenji Kurosawa, Hideyuki Tanabe, Nobuhiko Okamoto, Takumi Era, Hiroshi Sugiyama, Takahito Wada, Kohji Fukunaga
ZUSAMMENFASSUNG
Alpha-thalassemia X-linked intellectual disability (ATR-X) syndrome is caused by mutations in ATRX, which encodes a chromatin-remodeling protein. Genome-wide analyses in mouse and human cells indicate that ATRX tends to bind to G-rich sequences with a high potential to form G-quadruplexes. Here, we report that Atrx mutation induces aberrant upregulation of Xlr3b expression in the mouse brain, an outcome associated with neuronal pathogenesis displayed by ATR-X model mice. We show that ATRX normally binds to G-quadruplexes in CpG islands of the imprinted Xlr3b gene, regulating its expression by recruiting DNA methyltransferases. Xlr3b binds to dendritic mRNAs, and its overexpression inhibits dendritic transport of the mRNA encoding CaMKII-α, promoting synaptic dysfunction. Notably, treatment with 5-ALA, which is converted into G-quadruplex-binding metabolites, reduces RNA polymerase II recruitment and represses Xlr3b transcription in ATR-X model mice. 5-ALA treatment also rescues decreased synaptic plasticity and cognitive deficits seen in ATR-X model mice. Our findings suggest a potential therapeutic strategy to target G-quadruplexes and decrease cognitive impairment associated with ATR-X syndrome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
Anti-RNA-Polymerase II-Antikörper, Klon CTD4H8, clone CTD4H8, Upstate®, from mouse
Sigma-Aldrich
Monoklonaler Anti-Synaptophysin-Antikörper in Maus hergestellte Antikörper, clone SVP-38, ascites fluid