Merck
  • Development of evidence-based nursing-sensitive quality indicators for emergency nursing: A Delphi study.
Development of evidence-based nursing-sensitive quality indicators for emergency nursing: A Delphi study.
Journal of clinical nursing (2018-01-10)
Qiao-Yan Ju, Li-Hua Huang, Xue-Hong Zhao, Mei-Yuan Xing, Le-Wen Shao, Mei-Yun Zhang, Rong-Ya Shao
ZUSAMMENFASSUNG
To establish evidence-based nursing-sensitive quality indicators for emergency nursing in China. China lacks nursing-sensitive quality indicators necessary for assessing the quality of emergency nursing and essential to nursing management. Prospective. A literature search for relevant evidence-based studies was performed using several databases from January 2009-May 2014. Previously reported quality indicators were identified as appropriate for assessment by a panel of 40 experts in emergency medicine and nursing. Two successive rounds of Delphi surveys were conducted using questionnaires designed by the experts. Kendal's W coordination coefficients were calculated for indicator importance, rationality of calculation and feasibility of data collection. Thirty-three quality indicators were initially proposed for expert evaluation. After round 1 of expert discussion, Kendal's W coordination coefficients were .152 for importance, .092 for rationality and .141 for feasibility of data collection (all p < .001). Seven unsuitable items were discarded in round 1 and 11 discarded in round 2, which also added one new item. Finally, the experts reached consensus on 16 items established as appropriate nursing-sensitive quality indicators for emergency nursing care. Evidence-based nursing-sensitive quality indicators were established through a consensus of experts in emergency nursing and medicine. The current findings may provide a theoretical basis for establishing an emergency nursing quality database and improving the quality of emergency nursing care in China.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)