Merck
ZZ-dependent regulation of p62/SQSTM1 in autophagy.
Nature communications (2018-10-24)
Yi Zhang, Su Ran Mun, Juan F Linares, JaeWoo Ahn, Christina G Towers, Chang Hoon Ji, Brent E Fitzwalter, Michael R Holden, Wenyi Mi, Xiaobing Shi, Jorge Moscat, Andrew Thorburn, Maria T Diaz-Meco, Yong Tae Kwon, Tatiana G Kutateladze
ZUSAMMENFASSUNG
Autophagic receptor p62 is a critical mediator of cell detoxification, stress response, and metabolic programs and is commonly deregulated in human diseases. The diverse functions of p62 arise from its ability to interact with a large set of ligands, such as arginylated (Nt-R) substrates. Here, we describe the structural mechanism for selective recognition of Nt-R by the ZZ domain of p62 (p62ZZ). We show that binding of p62ZZ to Nt-R substrates stimulates p62 aggregation and macroautophagy and is required for autophagic targeting of p62. p62 is essential for mTORC1 activation in response to arginine, but it is not a direct sensor of free arginine in the mTORC1 pathway. We identified a regulatory linker (RL) region in p62 that binds p62ZZ in vitro and may modulate p62 function. Our findings shed new light on the mechanistic and functional significance of the major cytosolic adaptor protein p62 in two fundamental signaling pathways.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-LC3B in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
XIE62-1004, ≥98% (HPLC)