Merck
Alzheimer's-Causing Mutations Shift Aβ Length by Destabilizing γ-Secretase-Aβn Interactions.
Cell (2017-07-29)
Maria Szaruga, Bogdan Munteanu, Sam Lismont, Sarah Veugelen, Katrien Horré, Marc Mercken, Takaomi C Saido, Natalie S Ryan, Tatjana De Vos, Savvas N Savvides, Rodrigo Gallardo, Joost Schymkowitz, Frederic Rousseau, Nick C Fox, Carsten Hopf, Bart De Strooper, Lucía Chávez-Gutiérrez
ZUSAMMENFASSUNG
Alzheimer's disease (AD)-linked mutations in Presenilins (PSEN) and the amyloid precursor protein (APP) lead to production of longer amyloidogenic Aβ peptides. The shift in Aβ length is fundamental to the disease; however, the underlying mechanism remains elusive. Here, we show that substrate shortening progressively destabilizes the consecutive enzyme-substrate (E-S) complexes that characterize the sequential γ-secretase processing of APP. Remarkably, pathogenic PSEN or APP mutations further destabilize labile E-S complexes and thereby promote generation of longer Aβ peptides. Similarly, destabilization of wild-type E-S complexes by temperature, compounds, or detergent promotes release of amyloidogenic Aβ. In contrast, E-Aβn stabilizers increase γ-secretase processivity. Our work presents a unifying model for how PSEN or APP mutations enhance amyloidogenic Aβ production, suggests that environmental factors may increase AD risk, and provides the theoretical basis for the development of γ-secretase/substrate stabilizing compounds for the prevention of AD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® M2-Affinitätsgel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Presenilin-1-Antikörper, NT, Klon hPS1-NT, culture supernatant, clone hPS1-NT, Chemicon®
Sigma-Aldrich
GM 6001, InSolution, ≥95%, broad-spectrum inhibitor of MMPs
Sigma-Aldrich
γ-Sekretase-Inhibitor X, InSolution, ≥90%, 1 mM