Merck
Optimal protection against Salmonella infection requires noncirculating memory.
Proceedings of the National Academy of Sciences of the United States of America (2018-09-27)
Joseph M Benoun, Newton G Peres, Nancy Wang, Oanh H Pham, Victoria L Rudisill, Zachary N Fogassy, Paul G Whitney, Daniel Fernandez-Ruiz, Thomas Gebhardt, Quynh-Mai Pham, Lynn Puddington, Sammy Bedoui, Richard A Strugnell, Stephen J McSorley
ZUSAMMENFASSUNG
While CD4 Th1 cells are required for resistance to intramacrophage infections, adoptive transfer of Th1 cells is insufficient to protect against Salmonella infection. Using an epitope-tagged vaccine strain of Salmonella, we found that effective protection correlated with expanded Salmonella-specific memory CD4 T cells in circulation and nonlymphoid tissues. However, naive mice that previously shared a blood supply with vaccinated partners lacked T cell memory with characteristics of tissue residence and did not acquire robust protective immunity. Using a YFP-IFN-γ reporter system, we identified Th1 cells in the liver of immunized mice that displayed markers of tissue residence, including P2X7, ARTC2, LFA-1, and CD101. Adoptive transfer of liver memory cells after ARTC2 blockade increased protection against highly virulent bacteria. Taken together, these data demonstrate that noncirculating memory Th1 cells are a vital component of immunity to Salmonella infection and should be the focus of vaccine strategies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (20 °C)