Merck
  • The H3K4me3 histone demethylase Fbxl10 is a regulator of chemokine expression, cellular morphology, and the metabolome of fibroblasts.
The H3K4me3 histone demethylase Fbxl10 is a regulator of chemokine expression, cellular morphology, and the metabolome of fibroblasts.
The Journal of Biological Chemistry
Janzer, A; Stamm, K; Becker, A; Zimmer, A; Buettner, R; Kirfel, J
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Dimethyl-Histon-H3(Lys9)-Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Anti-Dimethyl-Histon H3 (Lys36) Antikörper, serum, Upstate®