Merck
  • ERAP1 promotes Hedgehog-dependent tumorigenesis by controlling USP47-mediated degradation of βTrCP.
ERAP1 promotes Hedgehog-dependent tumorigenesis by controlling USP47-mediated degradation of βTrCP.
Nature communications (2019-07-26)
Francesca Bufalieri, Paola Infante, Flavia Bernardi, Miriam Caimano, Paolo Romania, Marta Moretti, Ludovica Lospinoso Severini, Julie Talbot, Ombretta Melaiu, Mirella Tanori, Laura Di Magno, Diana Bellavia, Carlo Capalbo, Stéphanie Puget, Enrico De Smaele, Gianluca Canettieri, Daniele Guardavaccaro, Luca Busino, Angelo Peschiaroli, Simonetta Pazzaglia, Giuseppe Giannini, Gerry Melino, Franco Locatelli, Alberto Gulino, Olivier Ayrault, Doriana Fruci, Lucia Di Marcotullio
ZUSAMMENFASSUNG
The Hedgehog (Hh) pathway is essential for embryonic development and tissue homeostasis. Aberrant Hh signaling may occur in a wide range of human cancers, such as medulloblastoma, the most common brain malignancy in childhood. Here, we identify endoplasmic reticulum aminopeptidase 1 (ERAP1), a key regulator of innate and adaptive antitumor immune responses, as a previously unknown player in the Hh signaling pathway. We demonstrate that ERAP1 binds the deubiquitylase enzyme USP47, displaces the USP47-associated βTrCP, the substrate-receptor subunit of the SCFβTrCP ubiquitin ligase, and promotes βTrCP degradation. These events result in the modulation of Gli transcription factors, the final effectors of the Hh pathway, and the enhancement of Hh activity. Remarkably, genetic or pharmacological inhibition of ERAP1 suppresses Hh-dependent tumor growth in vitro and in vivo. Our findings unveil an unexpected role for ERAP1 in cancer and indicate ERAP1 as a promising therapeutic target for Hh-driven tumors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Peroxidase (HRP) in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse
Millipore
ANTI-FLAG® in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution