Merck
  • Sustained activation of P2X7 induces MMP-2-evoked cleavage and functional purinoceptor inhibition.
Sustained activation of P2X7 induces MMP-2-evoked cleavage and functional purinoceptor inhibition.
Journal of molecular cell biology (2017-10-11)
Christopher N J Young, Natalia Chira, Justyna Róg, Rasha Al-Khalidi, Magalie Benard, Ludovic Galas, Philippe Chan, David Vaudry, Krzysztof Zablocki, Dariusz C Górecki
ZUSAMMENFASSUNG
P2X7 purinoceptor promotes survival or cytotoxicity depending on extracellular adenosine triphosphate (ATP) stimulus intensity controlling its ion channel or P2X7-dependent large pore (LP) functions. Mechanisms governing this operational divergence and functional idiosyncrasy are ill-understood. We have discovered a feedback loop where sustained activation of P2X7 triggers release of active matrix metalloproteinase 2 (MMP-2), which halts ion channel and LP responses via the MMP-2-dependent receptor cleavage. This mechanism operates in cells as diverse as macrophages, dystrophic myoblasts, P2X7-transfected HEK293, and human tumour cells. Given that serum-born MMP-2 activity also blocked receptor functions, P2X7 responses in vivo may decrease in organs with permeable capillaries. Therefore, this mechanism represents an important fine-tuning of P2X7 functions, reliant on both cell-autonomous and extraneous factors. Indeed, it allowed evasion from the ATP-induced cytotoxicity in macrophages and human cancer cells with high P2X7 expression levels. Finally, we demonstrate that P2X7 ablation eliminated gelatinase activity in inflamed dystrophic muscles in vivo. Thus, P2X7 antagonists could be used as an alternative to highly toxic MMP inhibitors in treatments of inflammatory diseases and cancers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete ULTRA-Mini-Tabletten, EDTA-freier Proteasehemmer-Cocktail in EASYpacks, Tablets supplied in foil blister packs.
Sigma-Aldrich
Natriumorthovanadat, ≥90% (titration)
Sigma-Aldrich
Anti-Aktin, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-GAPDH in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution