Merck
Development of Intestinal Scaffolds that Mimic Native Mammalian Intestinal Tissue.
Tissue engineering. Part A (2019-01-18)
Mitchell R Ladd, Cait M Costello, Carolyn Gosztyla, Adam D Werts, Blake Johnson, William B Fulton, Laura Y Martin, Elizabeth J Redfield, Bryan Crawford, Rohan Panaparambil, Chhinder P Sodhi, John C March, David J Hackam
ZUSAMMENFASSUNG
This study is significant because it demonstrates an attempt to design a scaffold specifically for small intestine using a novel fabrication method, resulting in an architecture that resembles intestinal villi. In addition, we use the versatile polymer poly(glycerol sebacate) (PGS) for artificial intestine, which has tunable mechanical and degradation properties that can be harnessed for further fine-tuning of scaffold design. Moreover, the utilization of PGS allows for future development of growth factor and drug delivery from the scaffolds to promote artificial intestine formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Y-27632 -dihydrochlorid, ≥98% (HPLC)
Sigma-Aldrich
Pancreatin aus Schweinepankreas, 8 × USP specifications
Sigma-Aldrich
Gelatine aus Schweinehaut, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Anti--α-Glattmuskel-Aktin-Antikörper, Maus, monoklonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
4,4′-Methylen-bis(phenylisocyanat), 98%
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Bile extract porcine
Sigma-Aldrich
Poly(dimethylsiloxan), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Anti-Grün fluoreszierendes Protein-Antikörper, Chemicon®, from rabbit