Merck
  • Spatiotemporal material functionalization via competitive supramolecular complexation of avidin and biotin analogs.
Spatiotemporal material functionalization via competitive supramolecular complexation of avidin and biotin analogs.
Nature communications (2019-09-27)
Tom Kamperman, Michelle Koerselman, Cindy Kelder, Jan Hendriks, João F Crispim, Xandra de Peuter, Pieter J Dijkstra, Marcel Karperien, Jeroen Leijten
ZUSAMMENFASSUNG
Spatiotemporal control over engineered tissues is highly desirable for various biomedical applications as it emulates the dynamic behavior of natural tissues. Current spatiotemporal biomaterial functionalization approaches are based on cytotoxic, technically challenging, or non-scalable chemistries, which has hampered their widespread usage. Here we report a strategy to spatiotemporally functionalize (bio)materials based on competitive supramolecular complexation of avidin and biotin analogs. Specifically, an injectable hydrogel is orthogonally post-functionalized with desthiobiotinylated moieties using multivalent neutravidin. In situ exchange of desthiobiotin by biotin enables spatiotemporal material functionalization as demonstrated by the formation of long-range, conformal, and contra-directional biochemical gradients within complex-shaped 3D hydrogels. Temporal control over engineered tissue biochemistry is further demonstrated by timed presentation and sequestration of growth factors using desthiobiotinylated antibodies. The method's universality is confirmed by modifying hydrogels with biotinylated fluorophores, peptides, nanoparticles, enzymes, and antibodies. Overall, this work provides a facile, cytocompatible, and universal strategy to spatiotemporally functionalize materials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Triisopropylsilan, 98%
Sigma-Aldrich
Poly(dimethylsiloxan), viscosity 1.0 cSt (25 °C)
Desthiobiotin-iodoacetamide, ≥98% (HPLC), powder, pan-cysteine profiling tag