Merck
  • Interaction patches of procaspase-1 caspase recruitment domains (CARDs) are differently involved in procaspase-1 activation and receptor-interacting protein 2 (RIP2)-dependent nuclear factor κB signaling.
Interaction patches of procaspase-1 caspase recruitment domains (CARDs) are differently involved in procaspase-1 activation and receptor-interacting protein 2 (RIP2)-dependent nuclear factor κB signaling.
The Journal of biological chemistry (2011-08-25)
Kristof Kersse, Mohamed Lamkanfi, Mathieu J M Bertrand, Tom Vanden Berghe, Peter Vandenabeele
ZUSAMMENFASSUNG
Protein interaction domains belonging to the death domain-fold superfamily are six-helix bundles that mediate the assembly of large protein complexes involved in apoptotic and inflammatory signaling. Typically, death domains (DDs), a subfamily of the death domain-fold superfamily, harbor six delineated interaction patches on their surfaces that mediate three distinct and conserved types of interaction designated as types I, II, and III. Here, we show that caspase recruitment domains (CARDs), another subfamily of the death domain-fold superfamily, multimerize by employing at least two of the three reported interaction types that were identified in DDs. On the one hand, the CARD of procaspase-1 binds the apoptosis-associated speck-like protein containing a caspase recruitment domain (ASC) through a type I interaction that involves a patch surrounding residue Asp-27. On the other hand, the CARD of procaspase-1 auto-oligomerizes through a type III interaction involving a patch surrounding residue Arg-45. This oligomerization allows binding of receptor-interacting protein 2 (RIP2). In addition, we show that a 1:1 interaction between ASC and procaspase-1 is sufficient for procaspase-1 to gain proteolytic activity, whereas the formation of a higher order CARD complex involving ASC, procaspase-1, and RIP2 is required for effective procaspase-1-mediated NF-κB activation. These findings indicate that the CARD of procaspase-1 is differently involved in the formation of procaspase-1 activating platforms and procaspase-1-mediated, RIP2-dependent NF-κB activation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® M2-Affinitätsgel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Peroxidase (HRP) in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-VSV-G antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution