Merck
Host-Microbe-Drug-Nutrient Screen Identifies Bacterial Effectors of Metformin Therapy.
Cell (2019-09-03)
Rosina Pryor, Povilas Norvaisas, Georgios Marinos, Lena Best, Louise B Thingholm, Leonor M Quintaneiro, Wouter De Haes, Daniela Esser, Silvio Waschina, Celia Lujan, Reuben L Smith, Timothy A Scott, Daniel Martinez-Martinez, Orla Woodward, Kevin Bryson, Matthias Laudes, Wolfgang Lieb, Riekelt H Houtkooper, Andre Franke, Liesbet Temmerman, Ivana Bjedov, Helena M Cochemé, Christoph Kaleta, Filipe Cabreiro
ZUSAMMENFASSUNG
Metformin is the first-line therapy for treating type 2 diabetes and a promising anti-aging drug. We set out to address the fundamental question of how gut microbes and nutrition, key regulators of host physiology, affect the effects of metformin. Combining two tractable genetic models, the bacterium E. coli and the nematode C. elegans, we developed a high-throughput four-way screen to define the underlying host-microbe-drug-nutrient interactions. We show that microbes integrate cues from metformin and the diet through the phosphotransferase signaling pathway that converges on the transcriptional regulator Crp. A detailed experimental characterization of metformin effects downstream of Crp in combination with metabolic modeling of the microbiota in metformin-treated type 2 diabetic patients predicts the production of microbial agmatine, a regulator of metformin effects on host lipid metabolism and lifespan. Our high-throughput screening platform paves the way for identifying exploitable drug-nutrient-microbiome interactions to improve host health and longevity through targeted microbiome therapies. VIDEO ABSTRACT.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
L-Serin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Ribose, ≥99% (GC)
Sigma-Aldrich
GenElute Plasmid Miniprep-Kit, sufficient for 70 purifications
Sigma-Aldrich
Lithiumacetoacetat, ≥90% (HPLC)
Sigma-Aldrich
Indol-3-essigsäure Natriumsalz, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Anti-Maus-IgG (Fc-spezifische)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody
Sigma-Aldrich
Perhexilin -maleat (Salz), ≥98% (HPLC)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder