Merck
DCAF8, a novel MuRF1 interaction partner, promotes muscle atrophy.
Journal of cell science (2019-08-09)
Marcel Nowak, Benjamin Suenkel, Pablo Porras, Rebekka Migotti, Franziska Schmidt, Melanie Kny, Xiaoxi Zhu, Erich E Wanker, Gunnar Dittmar, Jens Fielitz, Thomas Sommer
ZUSAMMENFASSUNG
The muscle-specific RING-finger protein MuRF1 (also known as TRIM63) constitutes a bona fide ubiquitin ligase that routes proteins like several different myosin heavy chain proteins (MyHC) to proteasomal degradation during muscle atrophy. In two unbiased screens, we identified DCAF8 as a new MuRF1-binding partner. MuRF1 physically interacts with DCAF8 and both proteins localize to overlapping structures in muscle cells. Importantly, similar to what is seen for MuRF1, DCAF8 levels increase during atrophy, and the downregulation of either protein substantially impedes muscle wasting and MyHC degradation in C2C12 myotubes, a model system for muscle differentiation and atrophy. DCAF proteins typically serve as substrate receptors for cullin 4-type (Cul4) ubiquitin ligases (CRL), and we demonstrate that DCAF8 and MuRF1 associate with the subunits of such a protein complex. Because genetic downregulation of DCAF8 and inhibition of cullin activity also impair myotube atrophy in C2C12 cells, our data imply that the DCAF8 promotes muscle wasting by targeting proteins like MyHC as an integral substrate receptor of a Cul4A-containing ring ubiquitin ligase complex (CRL4A).This article has an associated First Person interview with the first author of the paper.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® M2-Affinitätsgel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Gelatine -Lösung, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody
Sigma-Aldrich
Anti-Glyceraldehyd-3-phosphat-dehydrogenase-Antikörper, Klon 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)-Peroxidase in Kaninchen hergestellte Antikörper, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoklonaler ANTI-HA-Antikörper in Maus hergestellte Antikörper, clone HA-7, ascites fluid
Sigma-Aldrich
Monoklonales Anti-c-Myc in Maus hergestellte Antikörper, clone 9E10, ascites fluid
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone V284, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Myc-Tag-Antikörper, Upstate®, from rabbit