Merck
  • Transcription-dependent targeting of Hda1C to hyperactive genes mediates H4-specific deacetylation in yeast.
Transcription-dependent targeting of Hda1C to hyperactive genes mediates H4-specific deacetylation in yeast.
Nature communications (2019-09-21)
So Dam Ha, Seokjin Ham, Min Young Kim, Ji Hyun Kim, Insoon Jang, Bo Bae Lee, Min Kyung Lee, Jin-Taek Hwang, Tae-Young Roh, TaeSoo Kim
ZUSAMMENFASSUNG
In yeast, Hda1 histone deacetylase complex (Hda1C) preferentially deacetylates histones H3 and H2B, and functionally interacts with Tup1 to repress transcription. However, previous studies identified global increases in histone H4 acetylation in cells lacking Hda1, a component of Hda1C. Here, we find that Hda1C binds to hyperactive genes, likely via the interaction between the Arb2 domain of Hda1 and RNA polymerase II. Additionally, we report that Hda1C specifically deacetylates H4, but not H3, at hyperactive genes to partially inhibit elongation. This role is contrast to that of the Set2-Rpd3S pathway deacetylating histones at infrequently transcribed genes. We also find that Hda1C deacetylates H3 at inactive genes to delay the kinetics of gene induction. Therefore, in addition to fine-tuning of transcriptional response via H3-specific deacetylation, Hda1C may modulate elongation by specifically deacetylating H4 at highly transcribed regions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Acetyl-Histon-H3-Antikörper, from rabbit
Sigma-Aldrich
Anti-Acetyl-Histon-H4-Antikörper, 1 mg/mL, Upstate®