Merck
Identification of PRDX6 as a regulator of ferroptosis.
Acta pharmacologica Sinica (2019-05-01)
Bin Lu, Xiao-Bing Chen, Yu-Cai Hong, Hong Zhu, Qiao-Jun He, Bo Yang, Mei-Dan Ying, Ji Cao
ZUSAMMENFASSUNG
Ferroptosis is a newly characterized iron-dependent form of nonapoptotic regulated cell death triggered by lipid reactive oxygen species (LOOH). The dysregulation of ferroptosis is highly related to cancer, and the induction of ferroptosis is also proposed as a potential strategy for cancer therapy. Although several key regulators have been identified that are involved in ferroptosis, the molecular mechanism underlying this process remains largely unknown. Here, we report that Peroxiredoxin-6 (PRDX6) is a bona fide negative regulator of ferroptotic cell death. The knockdown of intracellular PRDX6 significantly enhances LOOH and ferroptotic cell death triggered by ferroptosis inducers (Erastin and RSL-3), which is correlated with the transcriptional activation of heme oxygenase-1. Moreover, overexpression of heme oxygenase-1 enhances both Erastin- and RSL-3-triggered LOOH, suggesting that heme oxygenase-1 mediates PRDX6 silencing-enhanced ferroptosis. More importantly, the application of a specific PRDX6 phospholipase A2 (iPLA2) inhibitor, MJ-33, synergistically enhances the ferroptosis induced by Erastin, suggesting that PRDX6 removes LOOH through its iPLA2 activity. Thus, our findings reveal an essential role of PRDX6 in protecting cells against ferroptosis and provide a potential target to improve the antitumor activity of ferroptosis-based chemotherapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Doxycyclin -hyclat
Sigma-Aldrich
Deferoxamin -mesylat (Salz), powder, ≥92.5% (TLC)
Sigma-Aldrich
Sulforhodamin B Natriumsalz, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
MISSION® pLKO.1-puro Empty Vector Control Plasmid DNA, Contains no shRNA insert