Merck
  • Intranasal Tadalafil nanoemulsions: formulation, characterization and pharmacodynamic evaluation.
Intranasal Tadalafil nanoemulsions: formulation, characterization and pharmacodynamic evaluation.
Pharmaceutical development and technology (2019-06-20)
Bassant Elbardisy, Sally Galal, Doaa A Abdelmonsif, Nabila Boraie
ZUSAMMENFASSUNG
This study aims at improving the bioavailability of a poorly soluble phosphodiesterase-5 inhibitor; tadalafil (TD) via developing intranasal (IN) nanoemulsions (NEs). Optimum NE ingredients were selected based on solubility studies, emulsification tests, and phase diagram construction. Both o/w and w/o NEs were selected based on their drug loading capacity. Optimum formulations were subjected to physicochemical characterization and were assessed for nasal toxicity through biochemical analysis of tumor necrosis factor-α (TNF-α) and caspase-3 in rat nasal tissues. Pharmacodynamic study was performed via biochemical analysis of cyclic guanosine monophosphate (cGMP) in rat penis 2-h post-treatment and compared with oral suspension of Cialis® tablets. Optimum o/w and w/o NEs were successfully prepared using different ratios of Capmul-MCM-EP, Labrasol:Transcutol-HP (1:1) and water. Optimized formulations exhibited more than 4000-fold increase in TD solubility compared with its aqueous solubility. Both formulations were optically isotropic with the majority of globules in the nanometric-size range. Nasal toxicity study revealed no significant difference in values of TNF-α and caspase-3 between the NE-treated groups and the control group. Both TD-NEs succeeded to achieve a significant enhancement in cGMP levels. Our findings suggested that IN administration of the developed TD-NEs could provide a safe and effective alternative to TD oral delivery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Guanosin 3′,5′-zyklisches Monophosphat, ≥98% (HPLC), powder