Merck
Endocan regulates acute lung inflammation through control of leukocyte diapedesis.
Journal of applied physiology (Bethesda, Md. : 1985) (2019-07-12)
Alexandre Gaudet, Lucie Portier, Méline Prin, Marie-Christine Copin, Anne Tsicopoulos, Daniel Mathieu, Philippe Lassalle, Nathalie De Freitas Caires
ZUSAMMENFASSUNG
Acute respiratory distress syndrome is a severe form of respiratory failure, occurring in up to 20% of patients admitted to the intensive care unit with sepsis. Dysregulated leukocyte diapedesis is a major contributor to acute respiratory distress syndrome. Endocan is a circulating proteoglycan that binds to the leukocyte integrin leukocyte functional antigen-1 and blocks its interaction with its endothelial ligand, ICAM-1. The objective of this study was to evaluate the role of endocan in the control of acute lung inflammation. In vitro, endocan inhibited human leukocyte transendothelial migration as well as ICAM-1-dependent migration but had a very mild effect on ICAM-1-dependent adhesion. Endocan also acted as an inhibitor of transendothelial migration of mouse leukocytes. The effect of systemic administration of recombinant human endocan was assessed in a model of acute lung inflammation in BALB/c mice. Treatment with endocan 1 h after intratracheal LPS challenge reduced the alveolar inflammatory response, diminished histological features of acute lung injury, and improved respiratory function. These results highlight the anti-inflammatory role of human endocan and its protective effect against acute lung injury.NEW & NOTEWORTHY We show here that endocan inhibits ICAM-1-dependent human leukocyte transendothelial migration and ICAM-1-dependent adhesion. We also found that in BALB/c mice with tracheal LPS-induced acute lung injury treatment with recombinant human endocan reduces lung inflammation, notably through reduction of neutrophilic recruitment, and restores normal lung function. These results confirm the hypothesis that human endocan may have a protective effect against acute lung inflammation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Humanserum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Fibronectin-Humanplasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Endothelzellwachstumszusatz aus Rindernervengewebe, ECGS, suitable for cell culture
Sigma-Aldrich
Anti-Human IgG (Fc specific) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by trichloroacetic acid extraction
Sigma-Aldrich
Mouse MPO ELISA Kit, for plasma and cell culture supernatant
Sigma-Aldrich
Octoclothepin maleate salt, solid