Merck
Glucocorticoid feedback uncovers retrograde opioid signaling at hypothalamic synapses.
Nature neuroscience (2013-04-09)
Jaclyn I Wamsteeker Cusulin, Tamás Füzesi, Wataru Inoue, Jaideep S Bains
ZUSAMMENFASSUNG
Stressful experience initiates a neuroendocrine response culminating in the release of glucocorticoid hormones into the blood. Glucocorticoids feed back to the brain, causing adaptations that prevent excessive hormone responses to subsequent challenges. How these changes occur remains unknown. We found that glucocorticoid receptor activation in rodent hypothalamic neuroendocrine neurons following in vivo stress is a metaplastic signal that allows GABA synapses to undergo activity-dependent long-term depression (LTDGABA). LTDGABA was unmasked through glucocorticoid receptor-dependent inhibition of Regulator of G protein Signaling 4 (RGS4), which amplified signaling through postsynaptic metabotropic glutamate receptors. This drove somatodendritic opioid release, resulting in a persistent retrograde suppression of synaptic transmission through presynaptic μ receptors. Together, our data provide new evidence for retrograde opioid signaling at synapses in neuroendocrine circuits and represent a potential mechanism underlying glucocorticoid contributions to stress adaptation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mifepriston, ≥98%
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Naloxon -hydrochlorid Dihydrat, ≥98% (TLC and titration), powder