Merck
Molecular determinants of nephron vascular specialization in the kidney.
Nature communications (2019-12-15)
David M Barry, Elizabeth A McMillan, Balvir Kunar, Raphael Lis, Tuo Zhang, Tyler Lu, Edward Daniel, Masataka Yokoyama, Jesus M Gomez-Salinero, Angara Sureshbabu, Ondine Cleaver, Annarita Di Lorenzo, Mary E Choi, Jenny Xiang, David Redmond, Sina Y Rabbany, Thangamani Muthukumar, Shahin Rafii
ZUSAMMENFASSUNG
Although kidney parenchymal tissue can be generated in vitro, reconstructing the complex vasculature of the kidney remains a daunting task. The molecular pathways that specify and sustain functional, phenotypic and structural heterogeneity of the kidney vasculature are unknown. Here, we employ high-throughput bulk and single-cell RNA sequencing of the non-lymphatic endothelial cells (ECs) of the kidney to identify the molecular pathways that dictate vascular zonation from embryos to adulthood. We show that the kidney manifests vascular-specific signatures expressing defined transcription factors, ion channels, solute transporters, and angiocrine factors choreographing kidney functions. Notably, the ontology of the glomerulus coincides with induction of unique transcription factors, including Tbx3, Gata5, Prdm1, and Pbx1. Deletion of Tbx3 in ECs results in glomerular hypoplasia, microaneurysms and regressed fenestrations leading to fibrosis in subsets of glomeruli. Deciphering the molecular determinants of kidney vascular signatures lays the foundation for rebuilding nephrons and uncovering the pathogenesis of kidney disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Heparin Natriumsalz aus Schweinedarm, Grade I-A, ≥180 USP units/mg
Sigma-Aldrich
Medium 199, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-Kollagen-Antikörper, Typ IV, Chemicon®, from rabbit