Merck
  • ApoE (Apolipoprotein E) in Brain Pericytes Regulates Endothelial Function in an Isoform-Dependent Manner by Modulating Basement Membrane Components.
ApoE (Apolipoprotein E) in Brain Pericytes Regulates Endothelial Function in an Isoform-Dependent Manner by Modulating Basement Membrane Components.
Arteriosclerosis, thrombosis, and vascular biology (2019-11-02)
Yu Yamazaki, Mitsuru Shinohara, Akari Yamazaki, Yingxue Ren, Yan W Asmann, Takahisa Kanekiyo, Guojun Bu
ZUSAMMENFASSUNG
The ε4 allele of the APOE gene (APOE4) is the strongest genetic risk factor for Alzheimer disease when compared with the common ε3 allele. Although there has been significant progress in understanding how apoE4 (apolipoprotein E4) drives amyloid pathology, its effects on amyloid-independent pathways, in particular cerebrovascular integrity and function, are less clear. Approach and Results: Here, we show that brain pericytes, the mural cells of the capillary walls, differentially modulate endothelial cell phenotype in an apoE isoform-dependent manner. Extracellular matrix protein induction, tube-like structure formation, and barrier formation were lower with endothelial cells cocultured with pericytes isolated from apoE4-targeted replacement (TR) mice compared with those from apoE3-TR mice. Importantly, aged apoE4-targeted replacement mice had decreased extracellular matrix protein expression and increased plasma protein leakages compared with apoE3-TR mice. ApoE4 impairs pericyte-mediated basement membrane formation, potentially contributing to the cerebrovascular effects of apoE4.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Puromycin -dihydrochlorid aus Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Medium 199, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Insulin-Transferrin-Natriumselenit-Medienzusatz, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Albumin -Lösung, 22% in saline, contains azide, aseptically filled
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin-Flüssigsubstrat, besonders langsam, für ELISA, peroxidase substrate
Sigma-Aldrich
Anti-Kollagen-Antikörper, Typ IV, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Aquaporin-4-Antikörper, Chemicon®, from rabbit