Merck
  • ATRX In-Frame Fusion Neuroblastoma Is Sensitive to EZH2 Inhibition via Modulation of Neuronal Gene Signatures.
ATRX In-Frame Fusion Neuroblastoma Is Sensitive to EZH2 Inhibition via Modulation of Neuronal Gene Signatures.
Cancer cell (2019-10-22)
Zulekha A Qadeer, David Valle-Garcia, Dan Hasson, Zhen Sun, April Cook, Christie Nguyen, Aroa Soriano, Anqi Ma, Lyra M Griffiths, Maged Zeineldin, Dan Filipescu, Luz Jubierre, Asif Chowdhury, Orla Deevy, Xiang Chen, David B Finkelstein, Armita Bahrami, Elizabeth Stewart, Sara Federico, Soledad Gallego, Fumiko Dekio, Mary Fowkes, David Meni, John M Maris, William A Weiss, Stephen S Roberts, Nai-Kong V Cheung, Jian Jin, Miguel F Segura, Michael A Dyer, Emily Bernstein
ZUSAMMENFASSUNG
ATRX alterations occur at high frequency in neuroblastoma of adolescents and young adults. Particularly intriguing are the large N-terminal deletions of ATRX (Alpha Thalassemia/Mental Retardation, X-linked) that generate in-frame fusion (IFF) proteins devoid of key chromatin interaction domains, while retaining the SWI/SNF-like helicase region. We demonstrate that ATRX IFF proteins are redistributed from H3K9me3-enriched chromatin to promoters of active genes and identify REST as an ATRX IFF target whose activation promotes silencing of neuronal differentiation genes. We further show that ATRX IFF cells display sensitivity to EZH2 inhibitors, due to derepression of neurogenesis genes, including a subset of REST targets. Taken together, we demonstrate that ATRX structural alterations are not loss-of-function and put forward EZH2 inhibitors as a potential therapy for ATRX IFF neuroblastoma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-ATRX in Kaninchen hergestellte Antikörper, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Gelfiltrationsmarker-Kit für Proteine mit Molekülmassen von 29.000 bis 700.000 Da
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Target shRNA Control Plasmid DNA, Targets no known genes from any species
Sigma-Aldrich
Anti-REST-Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Anti-HP1α-Antikörper, Klon 15.19s2, clone 15.19s2, Upstate®, from mouse
Sigma-Aldrich
Anti-NXN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2