Merck
  • Myofibroblast-Specific TGFβ Receptor II Signaling in the Fibrotic Response to Cardiac Myosin Binding Protein C-Induced Cardiomyopathy.
Myofibroblast-Specific TGFβ Receptor II Signaling in the Fibrotic Response to Cardiac Myosin Binding Protein C-Induced Cardiomyopathy.
Circulation research (2018-12-20)
Qinghang Meng, Bidur Bhandary, Md Shenuarin Bhuiyan, Jeanne James, Hanna Osinska, Iñigo Valiente-Alandi, Kritton Shay-Winkler, James Gulick, Jeffery D Molkentin, Burns C Blaxall, Jeffrey Robbins
ZUSAMMENFASSUNG
Hypertrophic cardiomyopathy occurs with a frequency of about 1 in 500 people. Approximately 30% of those affected carry mutations within the gene encoding cMyBP-C (cardiac myosin binding protein C). Cardiac stress, as well as cMyBP-C mutations, can trigger production of a 40kDa truncated fragment derived from the amino terminus of cMyBP-C (Mybpc340kDa). Expression of the 40kDa fragment in mouse cardiomyocytes leads to hypertrophy, fibrosis, and heart failure. Here we use genetic approaches to establish a causal role for excessive myofibroblast activation in a slow, progressive genetic cardiomyopathy-one that is driven by a cardiomyocyte-intrinsic genetic perturbation that models an important human disease. TGFβ (transforming growth factor-β) signaling is implicated in a variety of fibrotic processes, and the goal of this study was to define the role of myofibroblast TGFβ signaling during chronic Mybpc340kDa expression. To specifically block TGFβ signaling only in the activated myofibroblasts in Mybpc340kDa transgenic mice and quadruple compound mutant mice were generated, in which the TGFβ receptor II (TβRII) alleles ( Tgfbr2) were ablated using the periostin ( Postn) allele, myofibroblast-specific, tamoxifen-inducible Cre ( Postnmcm) gene-targeted line. Tgfbr2 was ablated either early or late during pathological fibrosis. Early myofibroblast-specific Tgfbr2 ablation during the fibrotic response reduced cardiac fibrosis, alleviated cardiac hypertrophy, preserved cardiac function, and increased lifespan of the Mybpc340kDa transgenic mice. Tgfbr2 ablation late in the pathological process reduced cardiac fibrosis, preserved cardiac function, and prolonged Mybpc340kDa mouse survival but failed to reverse cardiac hypertrophy. Fibrosis and cardiac dysfunction induced by cardiomyocyte-specific expression of Mybpc340kDa were significantly decreased by Tgfbr2 ablation in the myofibroblast. Surprisingly, preexisting fibrosis was partially reversed if the gene was ablated subsequent to fibrotic deposition, suggesting that continued TGFβ signaling through the myofibroblasts was needed to maintain the heart fibrotic response to a chronic, disease-causing cardiomyocyte-only stimulus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti--α-Glattmuskel-Aktin-Antikörper, Maus, monoklonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Glyceraldehyd-3-phosphat-dehydrogenase-Antikörper, Klon 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-c-Myc-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Myc-Tag-Antikörper, Klon 9E10, clone 9E10, Upstate®, from mouse
Sigma-Aldrich
Anti-Troponin I Antibody, a.a. 186-192, clone C5, clone C5, Chemicon®, from mouse