Merck
  • Persistent Overexposure to N-Methyl-D-Aspartate (NMDA) Calcium-Dependently Downregulates Glutamine Synthetase, Aquaporin 4, and Kir4.1 Channel in Mouse Cortical Astrocytes.
Persistent Overexposure to N-Methyl-D-Aspartate (NMDA) Calcium-Dependently Downregulates Glutamine Synthetase, Aquaporin 4, and Kir4.1 Channel in Mouse Cortical Astrocytes.
Neurotoxicity research (2018-09-17)
Katarzyna Skowrońska, Marta Obara-Michlewska, Anna Czarnecka, Katarzyna Dąbrowska, Magdalena Zielińska, Jan Albrecht
ZUSAMMENFASSUNG
Astrocytes express N-methyl-D-aspartate (NMDA) receptor (NMDAR) but its functions in these cells are not well defined. This study shows that the sustained exposure (8-72 h) of mouse astrocytes to NMDA decreases the expression of the functional astroglia-specific proteins, glutamine synthetase (GS), and the water channel protein aquaporin-4 (AQP4) and also reduces GS activity. Similar to rat astrocytes (Obara-Michlewska et al. Neurochem Int 88:20-25, 2015), the exposure of mouse astrocytes to NMDA also decreased the expression of the inward rectifying potassium channel Kir4.1. NMDA failed to elicit the effects in those cells incubated in the absence of Ca2+ and in those in which the GluN1 subunit of the NMDAR was silenced with GluN1 siRNA. The downregulation of GS, AQP4, and Kir4.1 observed in vitro may reflect NMDAR-mediated alterations of astrocytic functions noted in central nervous system pathologies associated with increased glutamate (Glu) release and excitotoxic tissue damage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Gliafaserprotein-Antikörper, Klon GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Monoklonaler Anti-Neurofilament-200-Antikörper (phos. und nicht-phos.) in Maus hergestellte Antikörper, clone N52, ascites fluid
Sigma-Aldrich
Anti-NR1-Antikörper, CT, Upstate®, from mouse