Merck
  • Rat adrenomedullin induces a selective arterial vasodilation via CGRP1 receptors in the double-perfused mesenteric bed of the rat.
Rat adrenomedullin induces a selective arterial vasodilation via CGRP1 receptors in the double-perfused mesenteric bed of the rat.
Canadian journal of physiology and pharmacology (1995-07-01)
N Berthiaume, A Claing, H Lippton, A Cadieux, P D'Orléans-Juste
ZUSAMMENFASSUNG
The present work was undertaken to study the effect of rat adrenomedullin (rADM (1-50) and its C-terminal fragment (11-50)) in the endothelium-intact arterial and venous vasculatures of the rat perfused mesenteric bed. rADM (1-50) and the fragment rADM (11-50)(1-1000 pmol) induced a dose-dependent and endothelium-independent vasodilation on the arterial mesenteric vasculature. However, both peptides were inactive on the venous side of this vascular bed. The CGRP1 receptor antagonist, hCGRP8-37 (1 microM), markedly reduced the vasodilation caused by rADM (1-50) in the arterial mesenteric vasculature. Thus, our results show that rADM (1-50) in the arterial mesenteric vasculature. Thus, our results show that rADM (1-50) and its C-terminal fragment rADM(11-50) share properties similar to those of hCGRP. The blocking effect of hCGRP8-37 supports a role for CGRP1 receptor activation by adrenomedullin in this vascular preparation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adrenomedullin 11-50 rat