Merck
Antibody Therapy Targeting RAN Proteins Rescues C9 ALS/FTD Phenotypes in C9orf72 Mouse Model.
Neuron (2019-12-14)
Lien Nguyen, Fabio Montrasio, Amrutha Pattamatta, Solaleh Khoramian Tusi, Olgert Bardhi, Kevin D Meyer, Lindsey Hayes, Katsuya Nakamura, Monica Banez-Coronel, Alyssa Coyne, Shu Guo, Lauren A Laboissonniere, Yuanzheng Gu, Saravanakumar Narayanan, Benjamin Smith, Roger M Nitsch, Mark W Kankel, Mia Rushe, Jeffrey Rothstein, Tao Zu, Jan Grimm, Laura P W Ranum
ZUSAMMENFASSUNG
The intronic C9orf72 G4C2 expansion, the most common genetic cause of ALS and FTD, produces sense- and antisense-expansion RNAs and six dipeptide repeat-associated, non-ATG (RAN) proteins, but their roles in disease are unclear. We generated high-affinity human antibodies targeting GA or GP RAN proteins. These antibodies cross the blood-brain barrier and co-localize with intracellular RAN aggregates in C9-ALS/FTD BAC mice. In cells, α-GA1 interacts with TRIM21, and α-GA1 treatment reduced GA levels, increased GA turnover, and decreased RAN toxicity and co-aggregation of proteasome and autophagy proteins to GA aggregates. In C9-BAC mice, α-GA1 reduced GA as well as GP and GR proteins, improved behavioral deficits, decreased neuroinflammation and neurodegeneration, and increased survival. Glycosylation of the Fc region of α-GA1 is important for cell entry and efficacy. These data demonstrate that RAN proteins drive C9-ALS/FTD in C9-BAC transgenic mice and establish a novel therapeutic approach for C9orf72 ALS/FTD and other RAN-protein diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Duolink® In Situ Detektionsreagenzien Rot
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Kaninchen PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Maus MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Anti-Cholin-Acetyltransferase-Antikörper, Chemicon®, from goat
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glyceraldehyd-3-phosphat-dehydrogenase-Antikörper, Klon 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
(R)-MG132
Sigma-Aldrich
Tris[(1-Benzyl-1H-1,2,3-Triazol-4-yl)methyl]amin, 97%
Sigma-Aldrich
3-Methyladenin, autophagy inhibitor
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Sigma-Aldrich
Anti-V5 in Kaninchen hergestellte Antikörper, IgG fraction of antiserum