Merck
Epithelial Planar Bipolarity Emerges from Notch-Mediated Asymmetric Inhibition of Emx2.
Current biology : CB (2020-02-29)
Eva L Kozak, Subarna Palit, Jerónimo R Miranda-Rodríguez, Aleksandar Janjic, Anika Böttcher, Heiko Lickert, Wolfgang Enard, Fabian J Theis, Hernán López-Schier
ZUSAMMENFASSUNG
Most plane-polarized tissues are formed by identically oriented cells [1, 2]. A notable exception occurs in the vertebrate vestibular system and lateral-line neuromasts, where mechanosensory hair cells orient along a single axis but in opposite directions to generate bipolar epithelia [3-5]. In zebrafish neuromasts, pairs of hair cells arise from the division of a non-sensory progenitor [6, 7] and acquire opposing planar polarity via the asymmetric expression of the polarity-determinant transcription factor Emx2 [8-11]. Here, we reveal the initial symmetry-breaking step by decrypting the developmental trajectory of hair cells using single-cell RNA sequencing (scRNA-seq), diffusion pseudotime analysis, lineage tracing, and mutagenesis. We show that Emx2 is absent in non-sensory epithelial cells, begins expression in hair-cell progenitors, and is downregulated in one of the sibling hair cells via signaling through the Notch1a receptor. Analysis of Emx2-deficient specimens, in which every hair cell adopts an identical direction, indicates that Emx2 asymmetry does not result from auto-regulatory feedback. These data reveal a two-tiered mechanism by which the symmetric monodirectional ground state of the epithelium is inverted by deterministic initiation of Emx2 expression in hair-cell progenitors and a subsequent stochastic repression of Emx2 in one of the sibling hair cells breaks directional symmetry to establish planar bipolarity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Accutase® -Lösung, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Guanidin -hydrochlorid, Molecular Biology, ≥99%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Cas9-Protein, from Streptococcus pyogenes, recombinant, expressed in E. coli, 1X NLS