Merck
  • Advancing characterisation with statistics from correlative electron diffraction and X-ray spectroscopy, in the scanning electron microscope.
Advancing characterisation with statistics from correlative electron diffraction and X-ray spectroscopy, in the scanning electron microscope.
Ultramicroscopy (2020-01-31)
T P McAuliffe, A Foden, C Bilsland, D Daskalaki Mountanou, D Dye, T B Britton
ZUSAMMENFASSUNG
The routine and unique determination of minor phases in microstructures is critical to materials science. In metallurgy alone, applications include alloy and process development and the understanding of degradation in service. We develop a correlative method, exploring superalloy microstructures, which are examined in the scanning electron microscope (SEM) using simultaneous energy dispersive X-ray spectroscopy (EDS) and electron backscatter diffraction (EBSD). This is performed at an appropriate length scale for characterisation of carbide phases' shape, size, location, and distribution. EDS and EBSD data are generated using two different physical processes, but each provide a signature of the material interacting with the incoming electron beam. Recent advances in post-processing, driven by 'big data' approaches, include use of principal component analysis (PCA). Components are subsequently characterised to assign labels to a mapped region. To provide physically meaningful signals, the principal components may be rotated to control the distribution of variance. In this work, we develop this method further through a weighted PCA approach. We use the EDS and EBSD signals concurrently, thereby labelling each region using both EDS (chemistry) and EBSD (crystal structure) information. This provides a new method of amplifying signal-to-noise for very small phases in mapped regions, especially where the EDS or EBSD signal is not unique enough alone for classification.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)